• USA Home
  • Search Results

  • lithiumdodecylsulfate27233204456611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium dodecyl sulfate

3 Products
Synonym:   Dodecyl lithium sulfate, Dodecyl sulfate lithium salt, Lauryl sulfate lithium salt, Lithium lauryl sulfate
Image of Lithium dodecyl sulfate
CAS Number:
Linear Formula:
 CH3(CH2)11OSO3Li
Molecular Weight:
 272.33
 Beilstein/REAXYS Number:
 5186718
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
  Suitability
  Special Grade
  Product Line
  Molecular weight
  ID clarifier
  Assay
Show More