• USA Home
  • Search Results

  • lithiumhydride795758067811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium hydride

2 Products
Image of Lithium hydride
CAS Number:
Linear Formula:
 LiH
Molecular Weight:
 7.95
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More