• USA Home
  • Search Results

  • lithiumiodide133851037751211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium iodide

5 Products
Image of Lithium iodide
CAS Number:
Linear Formula:
 LiI
Molecular Weight:
 133.85
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Particle Dimension
  Physical Form
Show More