• USA Home
  • Search Results

  • lithiummetaborate49751345369511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium metaborate

4 Products
Synonym:   Boric acid lithium salt
Image of Lithium metaborate
CAS Number:
Linear Formula:
 LiBO2
Molecular Weight:
 49.75
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Loss
  Physical Form
Show More