• USA Home
  • Search Results

  • lithiummetaboratedihydrate85781529374011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium metaborate dihydrate

1 Products
Synonym:   Lithium borate (LiBO2 ) dihydrate
Image of Lithium metaborate dihydrate
CAS Number:
Linear Formula:
 LiBO2 · 2H2O
Molecular Weight:
 85.78
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More