• USA Home
  • Search Results

  • lithiumperchlorate10639779103911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium perchlorate

4 Products
Synonym:   Lithium cloricum, Perchloric acid, lithium salt
Image of Lithium perchlorate
CAS Number:
Linear Formula:
 LiClO4
Molecular Weight:
 106.39
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  UV-Absorption Values
  pH Value
  Physical Form
  Solubility
Show More