• USA Home
  • Search Results

  • lithiumsulfate109941037748711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium sulfate

3 Products
Synonym:   Dilithium sulfate
Image of Lithium sulfate
CAS Number:
Linear Formula:
 Li2SO4
Molecular Weight:
 109.94
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More