• USA Home
  • Search Results

  • lithiumtetrafluoroborate93751428307911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium tetrafluoroborate

4 Products
Synonym:   Lithium borofluoride, Lithium fluoroborate
Image of Lithium tetrafluoroborate
CAS Number:
Linear Formula:
 LiBF4
Molecular Weight:
 93.75
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Potency
Show More