• USA Home
  • Search Results

  • lithiumtrifluoromethanesulfonate156013345482911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium trifluoromethanesulfonate

2 Products
Synonym:   LiTf, Lithium triflate, Trifluoromethanesulfonic acid lithium salt
Image of Lithium trifluoromethanesulfonate
CAS Number:
Linear Formula:
 CF3SO3Li
Molecular Weight:
 156.01
 Beilstein/REAXYS Number:
 4301818
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More