• USA Home
  • Search Results

  • lithocholicacid3765743413911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithocholic acid

3 Products
Synonym:   3α-Hydroxy-5β-cholan-24-oic acid, 3α-Hydroxy-5β-cholanic acid, 5β-Cholan-24-oic acid-3α-ol
Image of Lithocholic acid
CAS Number:
Empirical Formula (Hill Notation):
 C24H40O3
Molecular Weight:
 376.57
 Beilstein/REAXYS Number:
 3217757
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Special Grade
  Manufacturer designation
Show More