• USA Home
  • Search Results

  • litholrubinebk42444528104911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithol Rubine BK

2 Products
Synonym:   3-Hydroxy-4-[(4-methyl-2-sulfophenyl)azo]-2-naphthoic acid calcium salt, E180
Image of Lithol Rubine BK
CAS Number:
Empirical Formula (Hill Notation):
 C18H12CaN2O6S
Molecular Weight:
 424.44
 Colour Index Number:
 15850
 E Number:
 E180
 EC Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More