• USA Home
  • Search Results

  • llacticdehydrogenasefromrabbitmuscle12345900160911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Lactic Dehydrogenase from rabbit muscle

2 Products
Synonym:   (S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD, Lactate
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 1.1.1.27  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Type
Show More