• USA Home
  • Search Results

  • llinalool1542512691011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Linalool

1 Products
Synonym:   (−)-Linalool, (R)-(−)-3,7-Dimethyl-1,6-octadien-3-ol
Image of <SC>L</SC>-Linalool
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 1721487
 Council of Europe no.:
 61
 EC Number:
 FEMA Number:
 2635
 Flavis number:
 2.013
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More