• USA Home
  • Search Results

  • llysine13c6hydrochloride18860122807786811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Lysine-13C6 hydrochloride

1 Products
Synonym:   (S)-2,6-Diaminocaproic acid-13C6, 13C Labeled Lys, 13C Labeled lysine, SILAC Amino Acid
Image of <SC>L</SC>-Lysine-<SUP>13</SUP>C<SUB>6</SUB> hydrochloride
CAS Number:
Linear Formula:
 H2N(13CH2)413CH(NH2)13CO2H · HCl
Molecular Weight:
 188.60
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More