• USA Home
  • Search Results

  • llysinemonohydrate164203966512811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Lysine monohydrate

2 Products
Image of <SC>L</SC>-Lysine monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C6H14N2O2 · H2O
Molecular Weight:
 164.20
 Beilstein/REAXYS Number:
 4349963
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More