• USA Home
  • Search Results

  • llysinemonohydrochloride1826565727211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Lysine monohydrochloride

8 Products
Synonym:   (S)-2,6-Diaminohexanoic acid monohydrochloride
Image of <SC>L</SC>-Lysine monohydrochloride
CAS Number:
Linear Formula:
 H2N(CH2)4CH(NH2)CO2H·HCl
Molecular Weight:
 182.65
 Beilstein/REAXYS Number:
 3563889
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Vapor pressure
  UV-Absorption Values
  Product Line
  Potency
  Melting Point
  Density
  Bulk Density
  Biological Source
  Analyte Suitability
  pH Value
  Physical Form
  Titertest
  Solubility
  Assay
Show More