• USA Home
  • Search Results

  • lmenthone154251407397311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Menthone

1 Products
Synonym:   (−)-Menthone, (1R,4S)-p-Menthan-3-one, (2S,5R)-2-Isopropyl-5-methylcyclohexanone, trans-menthone
Image of <SC>L</SC>-Menthone
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 2041368
 Council of Europe no.:
 2035
 EC Number:
 FEMA Number:
 2667
 Flavis number:
 7.176
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More