• USA Home
  • Search Results

  • lmethionine149216368311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Methionine

8 Products
Synonym:   (S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid
Image of <SC>L</SC>-Methionine
CAS Number:
Linear Formula:
 CH3SCH2CH2CH(NH2)CO2H
Molecular Weight:
 149.21
 Beilstein/REAXYS Number:
 1722294
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  UV-Absorption Values
  Manufacturer designation
  Biological Source
  Solubility
  Product Line
  Color
  Agency/Method
  Physical Form
  Special Grade
  Assay
Show More