• USA Home
  • Search Results

  • lmethioninesulfoximine180231598539411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Methionine sulfoximine

3 Products
Synonym:   L-S-(3-Amino-3-carboxypropyl)-S-methylsulfoximine
Image of <SC>L</SC>-Methionine sulfoximine
CAS Number:
Linear Formula:
 CH3S(O)(=NH)CH2CH2CH(NH2)CO2H
Molecular Weight:
 180.23
 Beilstein/REAXYS Number:
 6175446
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Package quantity
  Suitability
  Application(s)
  Product Line
  Assay
  Physical Form
Show More