• USA Home
  • Search Results

  • lorazepam3211684649111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Lorazepam

4 Products
Synonym:   7-Chloro-5-(2-chlorophenyl)-1,3-dihydro-3-hydroxy-2H-1,4-benzodiazepin-2-one
Image of (±)-Lorazepam
CAS Number:
Empirical Formula (Hill Notation):
 C15H10Cl2N2O2
Molecular Weight:
 321.16
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More