• USA Home
  • Search Results

  • lproline1151314785311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Proline

11 Products
Synonym:   (S)-Pyrrolidine-2-carboxylic acid
Image of <SC>L</SC>-Proline
CAS Number:
Empirical Formula (Hill Notation):
 C5H9NO2
Molecular Weight:
 115.13
 Beilstein/REAXYS Number:
 80810
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  UV-Absorption Values
  Potency
  Manufacturer designation
  Melting Point
  Loss
  Density
  Bulk Density
  Anal. Standard Form
  pH Value
  Product Line
  Biological Source
  Agency/Method
  Solubility
  Application(s)
  Color
  Physical Form
  Assay
Show More