• USA Home
  • Search Results

  • ltheanine17420308161611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Theanine

3 Products
Synonym:   Nγ-Ethyl-L-glutamine, L-Glutamic acid γ-(ethylamide), Suntheanine
Image of <SC>L</SC>-Theanine
CAS Number:
Empirical Formula (Hill Notation):
 C7H14N2O3
Molecular Weight:
 174.20
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More