• USA Home
  • Search Results

  • lthreonicacidhemicalciumsalt310277075361611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Threonic acid hemicalcium salt

1 Products
Synonym:   (2R,3S)-2,3,4-Trihydroxybutyric acid hemicalcium salt, L-Threonic acid calcium salt
Image of <SC>L</SC>-Threonic acid hemicalcium salt
CAS Number:
Empirical Formula (Hill Notation):
 C8H14CaO10
Molecular Weight:
 310.27
 Beilstein/REAXYS Number:
 5166008
 MDL number:
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More