• USA Home
  • Search Results

  • lthyroxine776875148911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Thyroxine

2 Products
Synonym:   3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4
Image of <SC>L</SC>-Thyroxine
CAS Number:
Linear Formula:
 HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
Molecular Weight:
 776.87
 Beilstein/REAXYS Number:
 2228515
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More