• USA Home
  • Search Results

  • ltryptophan204237322311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Tryptophan

7 Products
Synonym:   (S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid
Image of <SC>L</SC>-Tryptophan
CAS Number:
Empirical Formula (Hill Notation):
 C11H12N2O2
Molecular Weight:
 204.23
 Beilstein/REAXYS Number:
 86197
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Suitability
  Titertest
  Product Line
  Biological Source
  Analyte Suitability
  Physical Form
  Solubility
  Assay
Show More