• USA Home
  • Search Results

  • ltyrosine181196018411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Tyrosine

9 Products
Synonym:   (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine
Image of <SC>L</SC>-Tyrosine
CAS Number:
Linear Formula:
 4-(HO)C6H4CH2CH(NH2)CO2H
Molecular Weight:
 181.19
 Beilstein/REAXYS Number:
 392441
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Product Line
  Anal. Standard Form
  Analyte Suitability
  Suitability
  Biological Source
  Physical Form
  Solubility
  Assay
Show More