• USA Home
  • Search Results

  • ltyrosinemethylester19522108006411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Tyrosine methyl ester

1 Products
Image of <SC>L</SC>-Tyrosine methyl ester
CAS Number:
Linear Formula:
 4-(HO)C6H4CH2CH(NH2)COOCH3
Molecular Weight:
 195.22
 Beilstein/REAXYS Number:
 2372626
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More