• USA Home
  • Search Results

  • ltyrosinetertbutylester237291687412711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Tyrosine tert-butyl ester

1 Products
Image of <SC>L</SC>-Tyrosine <I>tert</I>-butyl ester
CAS Number:
Linear Formula:
 HOC6H4CH2CH(NH2)COOC(CH3)3
Molecular Weight:
 237.29
 Beilstein Registry Number:
 2696011
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More