• USA Home
  • Search Results

  • luciferasefromphotinuspyralisfirefly123456197000111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Luciferase from Photinus pyralis (firefly)

2 Products
Synonym:   Luciferase firefly
Image of Luciferase from <I>Photinus pyralis</I> (firefly)
CAS Number:
 Enzyme Commission (EC) Number:
 1.13.12.7  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Recombinant Host
  Ice Shipping
  Assay
Show More