• USA Home
  • Search Results

  • luminol1771652131311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Luminol

2 Products
Synonym:   3-Aminophthalhydrazide, 5-Amino-2,3-dihydro-1,4-phthalazinedione
Image of Luminol
CAS Number:
Empirical Formula (Hill Notation):
 C8H7N3O2
Molecular Weight:
 177.16
 Beilstein/REAXYS Number:
 383929
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More