• USA Home
  • Search Results

  • luteolin7glucoside44838537311511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Luteolin 7-glucoside

3 Products
Synonym:   3′,4′,5,7-Tetrahydroxyflavone 7-glucoside, Cynaroside, Glucoluteolin, Glucosylluteolin, Luteolin 7-O-β-D-glucoside, Luteoloside
Image of Luteolin 7-glucoside
CAS Number:
Empirical Formula (Hill Notation):
 C21H20O11
Molecular Weight:
 448.38
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
  Application(s)
  Manufacturer designation
Show More