• USA Home
  • Search Results

  • lvaline117157218411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Valine

10 Products
Synonym:   (S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid
Image of <SC>L</SC>-Valine
CAS Number:
Linear Formula:
 (CH3)2CHCH(NH2)CO2H
Molecular Weight:
 117.15
 Beilstein/REAXYS Number:
 1721136
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Anal. Standard Form
  Allergen
  Organoleptic property
  UV-Absorption Values
  Product Line
  Analyte Suitability
  Physical Form
  Titertest
  Biological Source
  Special Grade
  Solubility
  Assay
Show More