• USA Home
  • Search Results

  • lysineacetate206245728249211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lysine acetate

1 Products
Synonym:   (S)-2,6-Diaminohexanoic acid, L-Lysine acetate salt
Image of Lysine acetate
CAS Number:
Empirical Formula (Hill Notation):
 C6H14N2O2 · C2H4O2
Molecular Weight:
 206.24
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More