• USA Home
  • Search Results

  • magnesiumacetatetetrahydrate214451667478511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium acetate tetrahydrate

5 Products
Synonym:   Acetic acid magnesium salt
Image of Magnesium acetate tetrahydrate
CAS Number:
Linear Formula:
 (CH3COO)2Mg · 4H2O
Molecular Weight:
 214.45
 Beilstein/REAXYS Number:
 3730605
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
 eCl@ss:
 39021909
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  pH Value
  Solubility
  Product Line
  Special Grade
Show More