• USA Home
  • Search Results

  • magnesiumaluminatespinel142271206851811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium aluminate, spinel

1 Products
Synonym:   Magnesium aluminate
Image of Magnesium aluminate, spinel
CAS Number:
Linear Formula:
 MgO·Al2O3
Molecular Weight:
 142.27
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More