• USA Home
  • Search Results

  • magnesiumchloride9521778630311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium chloride

6 Products
Synonym:   Magnogene
Image of Magnesium chloride
CAS Number:
Linear Formula:
 MgCl2
Molecular Weight:
 95.21
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Solubility
  Product Line
  Particle Dimension
  Physical Form
Show More