• USA Home
  • Search Results

  • magnesiumchloridehexahydrate20330779118611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium chloride hexahydrate

17 Products
Image of Magnesium chloride hexahydrate
CAS Number:
Linear Formula:
 MgCl2 · 6H2O
Molecular Weight:
 203.30
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Restriction
  Physical Form
  Suitability
  UV-Absorption Condition
  Application(s)
  Agency/Method
  UV-Absorption Values
  Potency
  Melting Point
  Product Line
  pH Value
  Solubility
  Assay
  Special Grade
Show More

Product #
Description
M2670 Magnesium chloride hexahydrate, BioXtra, ≥99.0%,
M9272 Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%,
M2393 Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture,
63068 Magnesium chloride hexahydrate, BioUltra, for molecular biology, ≥99.0% (KT),
M0250 Magnesium chloride hexahydrate, ReagentPlus®, ≥99.0%,
1.05833 Magnesium chloride hexahydrate, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
13152 Magnesium chloride hexahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al,
M7304 Magnesium chloride hexahydrate, meets USP testing specifications,
1.05832 Magnesium chloride hexahydrate, cryst. EMPROVE® ESSENTIAL Ph Eur,BP,USP,JPC,FCC,E 511,
63064 Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT),
255777 Magnesium chloride hexahydrate, 99.995% trace metals basis,
1.37008 Magnesium chloride hexahydrate, cryst. EMPROVE® EXPERT Ph Eur,BP,USP,JPC,ACS,
1.02367 Magnesium chloride hexahydrate, cryst. EMPROVE® API Ph Eur,BP,JPC,USP,
1.01872 Magnesium chloride hexahydrate, CRYST. Ph Eur,JPC,USP,FCC,E 511,
1.72571 Magnesium chloride hexahydrate, extra pure, for table water FCC,E 511,
442615-M Magnesium chloride hexahydrate, ACS Reagent Grade.,
63033 Magnesium chloride hexahydrate, tested according to Ph Eur,