• USA Home
  • Search Results

  • magnesiumdgluconatehydrate41460anhydrousbasis363291511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium D-gluconate hydrate

2 Products
Synonym:   D-Gluconic acid hemimagnesium salt, D-Gluconic acid magnesium salt
Image of Magnesium <SC>D</SC>-gluconate hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C12H22MgO14 · xH2O
Molecular Weight:
 414.60 (anhydrous basis)
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More