• USA Home
  • Search Results

  • magnesiummethylcarbonatesolution13038486179411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium methyl carbonate solution

1 Products
Synonym:   MMC, Methoxymagnesium methyl carbonate, Methyl methoxymagnesium carbonate, Stile’s reagent
Image of Magnesium methyl carbonate solution
CAS Number:
Linear Formula:
 CH3OMgOCO2CH3
Molecular Weight:
 130.38
 Beilstein/REAXYS Number:
 4342967
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More