• USA Home
  • Search Results

  • magnesiumoxide4030130948411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium oxide

15 Products
Image of Magnesium oxide
CAS Number:
Linear Formula:
 MgO
Molecular Weight:
 40.30
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Semiconductor properties
  UV-Absorption Values
  Dielectric constant
  Composite Dimension
  pH Value
  Melting Point
  ID clarifier
  Density
  Restriction
  Agency/Method
  Transmittance
  Purity Grade
  Particle Dimension
  Bulk Density
  Loss
  Special Grade
  Physical Form
Show More

Product #
Description
529699 Magnesium oxide, ≥99.99% trace metals basis,
342793 Magnesium oxide, ≥99% trace metals basis, -325 mesh,
243388 Magnesium oxide, ACS reagent, 97%,
549649 Magnesium oxide, nanopowder, ≤50 nm particle size (BET),
307742 Magnesium oxide, light, 95%,
1.05865 Magnesium oxide, for analysis EMSURE®,
203718 Magnesium oxide, 99.99% trace metals basis,
63107 Magnesium oxide, tested according to Ph Eur, heavy,
220361 Magnesium oxide, -10-+50 mesh, 98%,
63089 Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT),
1.05862 Magnesium oxide, light extra pure Ph Eur,BP,USP,E 530,
13138 Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance),
634646 Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm,
1.05866 Magnesium oxide, for analysis (max.0.001% SO₄) EMSURE® ACS,
63090 Magnesium oxide, puriss. p.a., ACS reagent, ≥97% (calcined substance, KT),