• USA Home
  • Search Results

  • magnesiumstearate5912455704011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium stearate

3 Products
Synonym:   Stearic acid magnesium salt
Image of Magnesium stearate
CAS Number:
Linear Formula:
 [CH3(CH2)16CO2]2Mg
Molecular Weight:
 591.24
 Beilstein/REAXYS Number:
 3919702
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Special Grade
  Chemical Composition
  Solubility
Show More