• USA Home
  • Search Results

  • magnesiumsulfate12037748788911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium sulfate

7 Products
Image of Magnesium sulfate
CAS Number:
Empirical Formula (Hill Notation):
 MgSO4
Molecular Weight:
 120.37
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Application(s)
  Quality clarifier
  pH Value
  Product Line
  Physical Form
  Assay
Show More