• USA Home
  • Search Results

  • magnesiumsulfatehydrate12037anhydrousbasis2218908811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Magnesium sulfate hydrate

2 Products
Image of Magnesium sulfate hydrate
CAS Number:
Linear Formula:
 MgSO4 · aq
Molecular Weight:
 120.37 (anhydrous basis)
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More