• USA Home
  • Search Results

  • malachitegreenoxalatesalt46350243729811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Malachite Green oxalate salt

4 Products
Synonym:   N,N,N′,N′-Tetramethyl-4,4′-diaminotriphenylcarbenium oxalate, Basic Green 4
Image of Malachite Green oxalate salt
CAS Number:
Empirical Formula (Hill Notation):
 C23H25N2 · C2HO4 · 0.5C2H2O4
Molecular Weight:
 463.50
 Beilstein/REAXYS Number:
 3644550
 Colour Index Number:
 42000
 EC Number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Absorption
  Melting Point
  Special Grade
  Solubility
  Physical Form
Show More