• USA Home
  • Search Results

  • malathion3303612175511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Malathion

4 Products
Image of Malathion
CAS Number:
Empirical Formula (Hill Notation):
 C10H19O6PS2
Molecular Weight:
 330.36
 Beilstein/REAXYS Number:
 1804525
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Shelf Life
  Package quantity
  Manufacturer designation
  Special Grade
  Anal. Standard Form
  Physical Form
Show More