• USA Home
  • Search Results

  • maleicaciddisodiumsalthydrate16004anhydrousbasis2588069711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Maleic acid disodium salt hydrate

1 Products
Synonym:   Sodium maleate hydrate, Sodium toxilate hydrate
Image of Maleic acid disodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C4H2Na2O4 · xH2O
Molecular Weight:
 160.04 (anhydrous basis)
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More