• USA Home
  • Search Results

  • malonicacid1040614182211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Malonic acid

3 Products
Synonym:   Propanedioic acid
Image of Malonic acid
CAS Number:
Linear Formula:
 CH2(COOH)2
Molecular Weight:
 104.06
 Beilstein/REAXYS Number:
 1751370
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Density
  Application(s)
  Solubility
  Product Line
  Special Grade
  Assay
Show More