• USA Home
  • Search Results

  • maltitol3443158588611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Maltitol

4 Products
Synonym:   4-O-α-Glucopyranosyl-D-sorbitol
Image of Maltitol
CAS Number:
Empirical Formula (Hill Notation):
 C12H24O11
Molecular Weight:
 344.31
 Beilstein/REAXYS Number:
 89983
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Anal. Standard Form
Show More