• USA Home
  • Search Results

  • maltotriose50444110928011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Maltotriose

2 Products
Synonym:   α-D-Glc-(1→4)-α-D-Glc-(1→4)-D-Glc, O-α-DD-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose
Image of Maltotriose
CAS Number:
Empirical Formula (Hill Notation):
 C18H32O16
Molecular Weight:
 504.44
 Beilstein/REAXYS Number:
 1443481
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More