• USA Home
  • Search Results

  • maltylisobutyrate196206541614011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Maltyl isobutyrate

1 Products
Synonym:   Maltol isobutyrate
Image of Maltyl isobutyrate
CAS Number:
Empirical Formula (Hill Notation):
 C10H12O4
Molecular Weight:
 196.20
 Council of Europe no.:
 10739
 EC Number:
 FEMA Number:
 3462
 Flavis number:
 9.525
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Assay
Show More